(±)-1-Octen-4-ol ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B20707.06 B20707.14
B20707.06EA 78.5 EUR
B20707.06 B20707.14
(±)-1-Octen-4-ol ≥99%
(±)-1-Octen-4-ol

Purity: 99% CAS No.: 40575-42-6 Molecular Formula: C8H16O Formula Weight: 128.22 MDL No.: MFCD00021943

Formule: C₈H₁₆O
Dichtheid: 0,83
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00021943
CAS nummer: 40575-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR