4-Aminobutyraldehyde dimethyl acetal ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B21072.06 B21072.14
B21072.06EA 50.9 EUR
B21072.06 B21072.14
4-Aminobutyraldehyde dimethyl acetal ≥98%
4-Aminobutyraldehyde dimethyl acetal

Purity: 98+% CAS No.: 19060-15-2 Molecular Formula: C6H15NO2 Formula Weight: 133.19 Flash Point: 59°(138°F) Refractive Index: 1.4300 Boiling Point: 85°/22mm UN No.: 2734 MDL No.: MFCD00189373

Formule: C₆H₁₅NO₂
Kookpunt: 85 °C (22 mmHg)
Vlampunt: 59 °C (138 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00189373
CAS nummer: 19060-15-2
EINECS: 407-690-6
ADR: 8,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR