1-Naphthyl boronic acid ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B21219.03 B21219.06 B21219.14
B21219.03EA 41.6 EUR
B21219.03 B21219.06 B21219.14
1-Naphthyl boronic acid ≥97%
1-Naphthyl boronic acid

Purity: 97% CAS No.: 13922-41-3 Molecular Formula: C10H9BO2 Formula Weight: 171.99 Melting Point: 204-210° Beilstein Reference: 2937504

Formule: C₁₀H₉BO₂
Smeltpunt: 204…210 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00019722
CAS nummer: 13922-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR