Diisopropyl sulphide ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Isopropyl sulphide

B21302.09 B21302.18
B21302.09EA 43.1 EUR
B21302.09 B21302.18
Diisopropyl sulphide ≥99%
Diisopropyl sulphide

Purity: 99% CAS No.: 625-80-9 Molecular Formula: C6H14S Formula Weight: 118.24 Flash Point: 7°(44°F) Refractive Index: 1.4385 Odour: Stench Boiling Point: 119-120° Beilstein Reference: 1731257 UN No.: 1993 MDL No.: MFCD00008895

Formule: C₆H₁₄S
MW: 118.24 g/mol
Kookpunt: 119…120 °C
Smeltpunt: –69.1 °C
Dichtheid: 0.814
Vlampunt: 7 °C (44 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008895
CAS nummer: 625-80-9
EINECS: 210-911-7
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR