α,4-Dibromo-2-fluorotoluene ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B21331.06 B21331.14 B21331.22
B21331.06EA 42.7 EUR
B21331.06 B21331.14 B21331.22
α,4-Dibromo-2-fluorotoluene ≥98%
α,4-Dibromo-2-fluorotoluene

Purity: 98% CAS No.: 76283-09-5 Molecular Formula: C7H5Br2F Formula Weight: 267.92 Melting Point: 33-37° Boiling Point: 126°/9mm UN No.: 3261 MDL No.: MFCD00055467

Formule: C₇H₅Br₂F
Kookpunt: 126 °C (9 mmHg)
Smeltpunt: 33…37 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00055467
CAS nummer: 76283-09-5
EINECS: 278-412-7
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR