Thymol blue

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

B21370.09 B21370.18
B21370.09EA 23.8 EUR
B21370.09 B21370.18
Thymol blue
Thymol blue

CAS No.: 76-61-9 Molecular Formula: C27H30O5S Formula Weight: 466.60 Melting Point: 223° dec. Merck Reference: 14,9400 MDL No.: MFCD00005869

Formule: C₂₇H₃₀O₅S
MW: 466.6 g/mol
Smeltpunt: 223 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005869
CAS nummer: 76-61-9
EINECS: 200-973-3
Merck Index: 12,09541

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR