Pseudothiohydantoin 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B21461.06 B21461.14 B21461.22
B21461.06EA 35.7 EUR
B21461.06 B21461.14 B21461.22
Pseudothiohydantoin 97%
Pseudothiohydantoin

Purity: 97% CAS No.: 556-90-1 Molecular Formula: C3H4N2OS Formula Weight: 116.14 Melting Point: ca 240° dec. MDL No.: MFCD00003186

Formule: C₃H₄N₂OS
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003186
CAS nummer: 556-90-1
EINECS: 209-145-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR