2-Thiopheneethanol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B21562.06 B21562.14 B21562.22
B21562.06EA 38.4 EUR
B21562.06 B21562.14 B21562.22
2-Thiopheneethanol ≥98%
2-Thiopheneethanol

Purity: 98% CAS No.: 5402-55-1 Molecular Formula: C6H8OS Formula Weight: 128.19 Flash Point: 101°(213°F) Refractive Index: 1.5510 Boiling Point: 107-108°/12mm Beilstein Reference: 106985 MDL No.: MFCD00005462

Formule: C₆H₈OS
Kookpunt: 107…108 °C (12 mmHg)
Dichtheid: 1,150
Vlampunt: 101 °C (213 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005462
CAS nummer: 5402-55-1
EINECS: 226-452-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR