5-Bromo-m-xylene ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B21610.06 B21610.14 B21610.22
B21610.06EA 27.9 EUR
B21610.06 B21610.14 B21610.22
5-Bromo-m-xylene ≥98%
5-Bromo-m-xylene

Purity: 98% CAS No.: 556-96-7 Molecular Formula: C8H9Br Formula Weight: 185.07 Flash Point: 87°(188°F) Refractive Index: 1.5490 Boiling Point: 204° Beilstein Reference: 1926375 MDL No.: MFCD00000087

Formule: C₈H₉Br
Kookpunt: 204 °C
Dichtheid: 1,361
Vlampunt: 87 °C (188 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000087
CAS nummer: 556-96-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR