L(-)-Xylose ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B21622.06 B21622.14 B21622.22
B21622.06EA 44.9 EUR
B21622.06 B21622.14 B21622.22
L(-)-Xylose ≥99%
L(-)-Xylose

Purity: 99% CAS No.: 609-06-3 Molecular Formula: C5H10O5 Formula Weight: 150.13 Melting Point: 147-151° Beilstein Reference: 1723080

Formule: C₅H₁₀O₅
Kookpunt: 331 °C (1013 hPa)
Smeltpunt: 147…151 °C
Dichtheid: 1,525
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00151096
CAS nummer: 609-06-3
EINECS: 210-174-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR