(S)-(+)-1,2-Propanediol ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B21773.03 B21773.06 B21773.14
B21773.03EA 41.6 EUR
B21773.03 B21773.06 B21773.14
(S)-(+)-1,2-Propanediol ≥97%
(S)-(+)-1,2-Propanediol

Purity: 97% CAS No.: 4254-15-3 Molecular Formula: C3H8O2 Formula Weight: 76.10 Flash Point: 107°(224°F) Refractive Index: 1.4320 Boiling Point: 186-188° Beilstein Reference: 1718871 Merck Reference: 14,7855 Optical Rotation: +16° (neat) MDL No.: MFCD00004539

Formule: C₃H₈O₂
Kookpunt: 186…188 °C
Dichtheid: 1,040
Vlampunt: 107 °C (224 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004539
CAS nummer: 4254-15-3
Merck Index: 14,7855

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR