1-Iodoperfluorodecane ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Henicosafluoro-10-iododecane, Perfluorodecyl iodide, Perfluoro-1-iododecane

B21845.06 B21845.14
B21845.06EA 60.7 EUR
B21845.06 B21845.14
1-Iodoperfluorodecane ≥97%
1-Iodoperfluorodecane

Purity: 97% CAS No.: 423-62-1 Molecular Formula: CF3(CF2)9I Formula Weight: 645.98 Flash Point: None Melting Point: 63-67° Boiling Point: 195-200° Beilstein Reference: 1810506 MDL No.: MFCD00001065

Formule: C₁₀F₂₁I
MW: 645.98 g/mol
Kookpunt: 195…200 °C
Smeltpunt: 63…67 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001065
CAS nummer: 423-62-1
EINECS: 207-030-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR