Methyl-5-iodsalicylate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B22048.09 B22048.18
B22048.09EA 43.5 EUR
B22048.09 B22048.18
Methyl-5-iodsalicylate ≥99%
Methyl-5-iodsalicylate

Purity: 99% CAS No.: 4068-75-1 Molecular Formula: C8H7IO3 Formula Weight: 278.05 Melting Point: 75-78° MDL No.: MFCD00016462

Formule: C₈H₇IO₃
Smeltpunt: 75…78 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016462
CAS nummer: 4068-75-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR