4-tert-Butylbenzenesulphonyl chloride ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 4-tert-Butylbenzene-1-sulphonyl chloride

B22174.09 B22174.18
B22174.09EA 46.6 EUR
B22174.09 B22174.18
4-tert-Butylbenzenesulphonyl chloride ≥98%
4-tert-Butylbenzenesulphonyl chloride

Purity: 98% CAS No.: 15084-51-2 Molecular Formula: C10H13ClO2S Formula Weight: 232.73 Odour: Irritating Melting Point: 80-83° Beilstein Reference: 1104346 UN No.: 3261 MDL No.: MFCD00007449

Formule: C₁₀H₁₃ClO₂S
MW: 232.73 g/mol
Kookpunt: 165 °C (18 mmHg)
Smeltpunt: 80…83 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007449
CAS nummer: 15084-51-2
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR