Bis(2-mercaptoethyl)sulphide ≥90%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2,2'-Thiodiethanethiol, 2-Mercaptoethyl Sulphide

B22304.09 B22304.18 B22304.30
B22304.09EA 71.3 EUR
B22304.09 B22304.18 B22304.30
Bis(2-mercaptoethyl)sulphide ≥90%
Bis(2-mercaptoethyl)sulphide

Purity: 90+% CAS No.: 3570-55-6 Molecular Formula: HSCH2CH2SCH2CH2SH Formula Weight: 154.32 Flash Point: 90°(194°F) Boiling Point: 135-136°/10mm MDL No.: MFCD00004893

Formule: C₄H₁₀S₃
MW: 154.30 g/mol
Kookpunt: 135…136 °C (10 mmHg)
Dichtheid: 1.183
Vlampunt: 90 °C (194 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004893
CAS nummer: 3570-55-6
EINECS: 222-671-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR