Pentafluorothiophenol ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B22323.06 B22323.14 B22323.22
B22323.06EA 84.8 EUR
B22323.06 B22323.14 B22323.22
Pentafluorothiophenol ≥97%
Pentafluorothiophenol

Purity: 97% CAS No.: 771-62-0 Molecular Formula: C6HF5S Formula Weight: 200.13 Flash Point: 51°(123°F) Refractive Index: 1.4650 Odour: Stench Melting Point: -24° Boiling Point: 143° Beilstein Reference: 1876292 UN No.: 2920 MDL No.: MFCD00004828

Formule: C₆HF₅S
Kookpunt: 143 °C
Smeltpunt: –24 °C
Dichtheid: 1,572
Vlampunt: 51 °C (123 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004828
CAS nummer: 771-62-0
EINECS: 212-236-3
UN: 2920
ADR: 8,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR