N-Fmoc-L-norleucine ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B22475.03 B22475.06 B22475.14
B22475.03EA 23.2 EUR
B22475.03 B22475.06 B22475.14
N-Fmoc-L-norleucine ≥98%
N-Fmoc-L-norleucine

Purity: 98% CAS No.: 77284-32-3 Molecular Formula: C21H23NO4 Formula Weight: 353.42 Storage Temperature: 2°C to 8°C Melting Point: 140-142° MDL No.: MFCD00037537

Formule: C₂₁H₂₃NO₄
Smeltpunt: 140…142 °C
MDL Nummer: MFCD00037537
CAS nummer: 77284-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR