(±)-2-Bromopropionyl bromide ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B22504.22 B22504.36
B22504.22EA 38.9 EUR
B22504.22 B22504.36
(±)-2-Bromopropionyl bromide ≥97%
(±)-2-Bromopropionyl bromide

Purity: 97% CAS No.: 563-76-8 Molecular Formula: CH3CHBrCOBr Formula Weight: 215.88 Flash Point: 110°(230°F) Refractive Index: 1.5210 Boiling Point: 48-50°/10mm Beilstein Reference: 1071331 UN No.: 3265 MDL No.: MFCD00000142

Formule: C₃H₄Br₂O
Kookpunt: 48…50 °C (10 mmHg)
Dichtheid: 2,061
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000142
CAS nummer: 563-76-8
EINECS: 209-261-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR