Allyl methyl sulphide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Allyl(methyl)sulphane

B22514.06 B22514.14
B22514.06EA 24.5 EUR
B22514.06 B22514.14
Allyl methyl sulphide ≥98%
Allyl methyl sulphide

Purity: 98% CAS No.: 10152-76-8 Molecular Formula: CH2=CHCH2SCH3 Formula Weight: 88.17 Storage Temperature: 2°C to 8°C Flash Point: 18°(65°F) Refractive Index: 1.4714 Odour: Stench Boiling Point: 91-93° UN No.: 1993 MDL No.: MFCD00008657

Formule: C₄H₈S
MW: 88.17 g/mol
Kookpunt: 91…93 °C
Dichtheid: 0.803
Vlampunt: 18 °C (65 °F)
MDL Nummer: MFCD00008657
CAS nummer: 10152-76-8
EINECS: 233-422-0
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR