(Methylthio)acetic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B22531.14 B22531.03 B22531.06
B22531.14EA 225 EUR
B22531.14 B22531.03 B22531.06
(Methylthio)acetic acid ≥98%
(Methylthio)acetic acid

Purity: 98% CAS No.: 2444-37-3 Molecular Formula: CH3SCH2CO2H Formula Weight: 106.15 Flash Point: 110°(230°F) Refractive Index: 1.4950 Odour: Stench Melting Point: 13-14° Boiling Point: 99-100°/4mm Beilstein Reference: 1740201 MDL No.: MFCD00075444

Formule: C₃H₆O₂S
Kookpunt: 99…100 °C (4 mmHg)
Smeltpunt: 13…14 °C
Dichtheid: 1,224
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00075444
CAS nummer: 2444-37-3
EINECS: 219-483-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR