1-Bromo-2,4-dinitrobenzene ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B22867.14 B22867.22
B22867.14EA 77.3 EUR
B22867.14 B22867.22
1-Bromo-2,4-dinitrobenzene ≥98%
1-Bromo-2,4-dinitrobenzene

Purity: 98% CAS No.: 584-48-5 Molecular Formula: C6H3BrN2O4 Formula Weight: 247.01 Melting Point: 70-73° UN No.: 3459 MDL No.: MFCD00041873

Formule: C₆H₃BrN₂O₄
Smeltpunt: 70…73 °C
Dichtheid: >1 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041873
CAS nummer: 584-48-5
EINECS: 209-539-8
UN: 3459
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR