Creatinine ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B23097.09 B23097.18 B23097.30
B23097.09EA 26.3 EUR
B23097.09 B23097.18 B23097.30
Creatinine ≥98%
Creatinine

Purity: 98% CAS No.: 60-27-5 Molecular Formula: C4H7N3O Formula Weight: 113.12 Odour: Odourless pH: 7.5-8.5 Melting Point: ca 255° dec. Beilstein Reference: 112061 Merck Reference: 14,2569 MDL No.: MFCD00059730

Formule: C₄H₇N₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00059730
CAS nummer: 60-27-5
EINECS: 200-466-7
Merck Index: 14,2569

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR