3-(Hydroxymethyl)benzyl alcohol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B23417.03 B23417.06 B23417.14
B23417.03EA 28.6 EUR
B23417.03 B23417.06 B23417.14
3-(Hydroxymethyl)benzyl alcohol ≥98%
3-(Hydroxymethyl)benzyl alcohol

Purity: 98% CAS No.: 626-18-6 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 56-60° Boiling Point: 154-159°/13mm MDL No.: MFCD00004649

Formule: C₈H₁₀O₂
Kookpunt: 154…159 °C (13 mmHg)
Smeltpunt: 56…60 °C
Dichtheid: 1,161 g/cm³ (18 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004649
CAS nummer: 626-18-6
EINECS: 210-934-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR