2-Ethylhexyl bromide 96% stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Stabilisatie: Stabilised with copper

B23532.14 B23532.22
B23532.14EA 30.7 EUR
B23532.14 B23532.22
2-Ethylhexyl bromide 96% stabilised
2-Ethylhexyl bromide

Stabilized with copper


Purity: 96% CAS No.: 18908-66-2 Molecular Formula: C8H17Br Formula Weight: 193.13 Storage Temperature: 2°C to 8°C Flash Point: 69°(156°F) Refractive Index: 1.4538 Boiling Point: 94-95°/35mm Beilstein Reference: 1098313 MDL No.: MFCD00000220

Formule: C₈H₁₇Br
Kookpunt: 94…95 °C (35 mmHg)
Dichtheid: 1,086
Vlampunt: 69 °C (156 °F)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000220
CAS nummer: 18908-66-2
EINECS: 242-659-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR