4-Biphenylboronic acid ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B23703.03 B23703.06 B23703.14
B23703.03EA 33.6 EUR
B23703.03 B23703.06 B23703.14
4-Biphenylboronic acid ≥97%
4-Biphenylboronic acid

Purity: 98+% CAS No.: 5122-94-1 Molecular Formula: C12H11BO2 Formula Weight: 198.03 Odour: Odourless Melting Point: 238-247° Beilstein Reference: 2937410 MDL No.: MFCD00093311

Formule: C₁₂H₁₁BO₂
Smeltpunt: 238…247 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00093311
CAS nummer: 5122-94-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR