Nitro blue tetrazolium chloride (NBT) tech. 90%, Technical Grade

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

B23792.02 B23792.88
B23792.02EA 83.1 EUR
B23792.02 B23792.88
Nitro blue tetrazolium chloride (NBT) tech. 90%, Technical Grade
Nitro blue tetrazolium chloride (NBT)

Purity: 90% CAS No.: 298-83-9 Molecular Formula: C40H30Cl2N10O6 Formula Weight: 817.65 Melting Point: ca 190° dec. Beilstein Reference: 4115923

Formule: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Smeltpunt: ∼ 190 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012159
CAS nummer: 298-83-9
EINECS: 206-067-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR