5-Aminosalicylic acid 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B23970.22 B23970.36
B23970.22EA 53.3 EUR
B23970.22 B23970.36
5-Aminosalicylic acid 95%
5-Aminosalicylic acid

Purity: 95% CAS No.: 89-57-6 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: ca 280° dec. Beilstein Reference: 2090421 Merck Reference: 14,5904 MDL No.: MFCD00007877

Formule: C₇H₇NO₃
Kookpunt: 347 °C (1013 hPa)
Dichtheid: 1,568 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007877
CAS nummer: 89-57-6
EINECS: 201-919-1
Merck Index: 14,5904

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR