Tetrabutylammonium acetate 1.0 M in aqueous solution

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Tetra-n-butylammonium acetate

B24773.14 B24773.22
B24773.14EA 53.1 EUR
B24773.14 B24773.22
Tetrabutylammonium acetate 1.0 M in aqueous solution
Tetrabutylammonium acetate

CAS No.: 10534-59-5 Molecular Formula: C18H39NO2 Formula Weight: 301.51 Refractive Index: 1.3890 Boiling Point: 100° Beilstein Reference: 3599376

Formule: C₁₈H₃₉NO₂
MW: 301.51 g/mol
Kookpunt: 100 °C
Dichtheid: 0.990
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00043208
CAS nummer: 10534-59-5
EINECS: 234-101-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR