Sodium-2-ethylhexanoate ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B24960.0B B24960.0I B24960.30
B24960.0BEA 94.8 EUR
B24960.0B B24960.0I B24960.30
Sodium-2-ethylhexanoate ≥97%
Sodium-2-ethylhexanoate

Purity: 97% CAS No.: 19766-89-3 Molecular Formula: C8H15NaO2 Formula Weight: 166.20 Odour: Odourless Melting Point: 300° MDL No.: MFCD00014007

Formule: C₈H₁₅NaO₂
Smeltpunt: >300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00014007
CAS nummer: 19766-89-3
EINECS: 243-283-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR