Pinacol ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B25006.14 B25006.22 B25006.36
B25006.14EA 47 EUR
B25006.14 B25006.22 B25006.36
Pinacol ≥99%
Pinacol

Purity: 99% CAS No.: 76-09-5 Molecular Formula: C6H14O2 Formula Weight: 118.18 Flash Point: 77°(170°F) Melting Point: 42-45° Boiling Point: 171-172° Beilstein Reference: 1340501 Merck Reference: 14,7439 MDL No.: MFCD00004462

Formule: C₆H₁₄O₂
Kookpunt: 171…172 °C
Smeltpunt: 42…45 °C
Dichtheid: 0,967 g/cm³ (20 °C)
Vlampunt: 77 °C (170 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004462
CAS nummer: 76-09-5
EINECS: 200-933-5
Merck Index: 14,7439

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR