5-Isoquinolinol tech. 90%, Technical Grade

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B25052.14
B25052.14EA 0 EUR
B25052.14
5-Isoquinolinol tech. 90%, Technical Grade
5-Isoquinolinol

Purity: 90% CAS No.: 2439-04-5 Molecular Formula: C9H7NO Formula Weight: 145.16 MDL No.: MFCD00006906

Formule: C₉H₇NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006906
CAS nummer: 2439-04-5
EINECS: 219-456-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR