EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B25057.14 B25057.22 B25057.06
B25057.14EA 265 EUR
B25057.14 B25057.22 B25057.06
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide)

Purity: 98% CAS No.: 1892-57-5 Molecular Formula: C8H17N3 Formula Weight: 155.24 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Refractive Index: 1.4610 Boiling Point: 66-68°/1mm Beilstein Reference: 507429 UN No.: 2735 MDL No.: MFCD00044916

Formule: C₈H₁₇N₃
Kookpunt: 66…68 °C (1 mmHg)
Smeltpunt: 114…115 °C
Dichtheid: 0,885
Vlampunt: >110 °C (230 °F)
MDL Nummer: MFCD00044916
CAS nummer: 1892-57-5
EINECS: 217-579-2
ADR: 8,III

Order Now

Specification Test Results

Appearance (Color) Clear colorless to yellow
Form Liquid
Identification (FTIR) Conforms
Refractive Index 1.4590-1.4630 @ 20°C
Assay (GC) ≥96.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR