Itaconic anhydride ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Methylenesuccinic anhydride, 3-Methylenedihydrofuran-2,5-dione

B25216.14 B25216.22 B25216.36
B25216.14EA 48.7 EUR
B25216.14 B25216.22 B25216.36
Itaconic anhydride ≥97%
Itaconic anhydride

Purity: 97% CAS No.: 2170-03-8 Molecular Formula: C5H4O3 Formula Weight: 112.09 Melting Point: 68-72° Boiling Point: 114-115°/12mm MDL No.: MFCD00005530

Formule: C₅H₄O₃
MW: 112.08 g/mol
Kookpunt: 114…115 °C (12 mmHg)
Smeltpunt: 68…72 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005530
CAS nummer: 2170-03-8
EINECS: 218-518-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR