2-Amino-3,4-dimethylbenzoic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B25313.06 B25313.14
B25313.06EA 83.9 EUR
B25313.06 B25313.14
2-Amino-3,4-dimethylbenzoic acid ≥98%
2-Amino-3,4-dimethylbenzoic acid

Purity: 98+% CAS No.: 50419-58-4 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Melting Point: ca 176° dec. MDL No.: MFCD00130041

Formule: C₉H₁₁NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00130041
CAS nummer: 50419-58-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR