2-Bromo-5-chloroanisole ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 1-Bromo-4-chloro-2-methoxybenzene

B25512.06 B25512.14
B25512.06EA 93.4 EUR
B25512.06 B25512.14
2-Bromo-5-chloroanisole ≥98%
2-Bromo-5-chloroanisole

Purity: 98+% CAS No.: 174913-09-8 Molecular Formula: C7H6BrClO Formula Weight: 221.48 Refractive Index: 1.5875

Formule: C₇H₆BrClO
MW: 221.48 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03790889
CAS nummer: 174913-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR