5-Bromo-2-(trifluoromethyl)pyridine ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B25731.03 B25731.06
B25731.03EA 89.1 EUR
B25731.03 B25731.06
5-Bromo-2-(trifluoromethyl)pyridine ≥97%
5-Bromo-2-(trifluoromethyl)pyridine

Purity: 97% CAS No.: 436799-32-5 Molecular Formula: C6H3BrF3N Formula Weight: 226.00 Melting Point: 39-40° Boiling Point: 76-78°/18mm MDL No.: MFCD06657686

Formule: C₆H₃BrF₃N
Kookpunt: 76…78 °C (18 mmHg)
Smeltpunt: 39…40 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06657686
CAS nummer: 436799-32-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR