L-Methionine methyl ester hydrochloride

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4000448.0025 4000448.0100
BACE4000448.0025EA 0 EUR
BACE4000448.0025 BACE4000448.0100
L-Methionine methyl ester hydrochloride
L-Methionine methyl ester hydrochloride
Formule: C₆H₁₃NO₂S·HCl
MDL Nummer: MFCD00012491
CAS nummer: 2491-18-1
EINECS: 219-651-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR