Boc-D-Pro-OH

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4000664.0005 4000664.0025
BACE4000664.0005EA 0 EUR
BACE4000664.0005 BACE4000664.0025
Boc-D-Pro-OH
Boc-D-Pro-OH
Formule: C₁₀H₁₇NO₄
MDL Nummer: MFCD00063226
CAS nummer: 37784-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR