N-Fmoc-glycine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetic acid, Fmoc-Gly-OH

4003078.0025 4003078.0100 B-1330.1000 4003078.0500
BACE4003078.0025EA 0 EUR
BACE4003078.0025 BACE4003078.0100 BACEB-1330.1000 BACE4003078.0500
N-Fmoc-glycine
N-Fmoc-glycine
Formule: C₁₇H₁₅NO₄
MW: 297.3 g/mol
MDL Nummer: MFCD00037140
CAS nummer: 29022-11-5
EINECS: 249-373-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR