4-Bromo-DL-phenylalanine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4007001.0005 4007001.0025
BACE4007001.0005EA 0 EUR
BACE4007001.0005 BACE4007001.0025
4-Bromo-DL-phenylalanine
4-Bromo-DL-phenylalanine
Formule: C₉H₁₀BrNO₂
MDL Nummer: MFCD00002599
CAS nummer: 14091-15-7
EINECS: 237-941-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR