L(+)-Penicillamine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4031087.0005 4031087.0025
BACE4031087.0005EA 0 EUR
BACE4031087.0005 BACE4031087.0025
L(+)-Penicillamine
L(+)-Penicillamine
Formule: C₅H₁₁NO₂S
MDL Nummer: MFCD00064303
CAS nummer: 1113-41-3
EINECS: 214-203-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR