D(-)-Ornithine hydrochloride

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4031134.0005 4031134.0025 4031134.0100
BACE4031134.0005EA 0 EUR
BACE4031134.0005 BACE4031134.0025 BACE4031134.0100
D(-)-Ornithine hydrochloride
D-(-)-Ornithine hydrochloride
Formule: C₅H₁₂N₂O₂·HCl
MDL Nummer: MFCD00012917
CAS nummer: 16682-12-5
EINECS: 240-729-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR