N-Fmoc-L-valine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B-1455.0025 B-1455.0100 B-1455.1000 B-1455.0500
BACEB-1455.0025EA 70.08 EUR
BACEB-1455.0025 BACEB-1455.0100 BACEB-1455.1000 BACEB-1455.0500
N-Fmoc-L-valine
N-Fmoc-L-valine
Formule: C₂₀H₂₁NO₄
MDL Nummer: MFCD00037124
CAS nummer: 68858-20-8
EINECS: 272-515-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR