Ultrasonic generator, GM 5200
Leverancier: BANDELIN-ELECTRONIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3736
BAND3736EA 3290 EUR
BAND3736
Ultrasonic generator, GM 5200
Homogenisatoren Dispergeerstaven
There are two different ultrasonic generators to choose from. They are specified for various applications due to their different power ranges. The easy-care and robust plastic housing, equipped with a practical recessed grip for easy transportation and positioning on the laboratory bench, is identical for both versions.
The various SONOPULS ultrasonic homogenisers in the HD 5000 series can be connected directly to the ultrasonic generators depending on their compatibility. Mains connection 230 VAC (±10 %), 50/60 Hz.
The modern 7" touch display offers intuitive, user-friendly operation.More Product Information