Ultrasonic converter, HD 5000
Leverancier: BANDELIN-ELECTRONIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3749
BAND3749EA 1570 EUR
BAND3749
Ultrasonic converter, HD 5000
Homogenisatoren Ultrasone celscheiders
An ultrasonic converter is used to convert the electrical energy supplied by the ultrasonic generator into mechanical vibrations.
Ultrasonic operation can be started and stopped by operating the 'Start', 'Pause' and 'Stop' buttons on the touch display or via the button on the ultrasonic converter.
With the exception of the UW 5020, which operates at 30 kHz, all other SONOPULS ultrasonic converters in the 5000 series operate at an ultrasonic frequency of 20 kHz.
There is a button on the ultrasonic converter. This can be used to switch ultrasonic operation ON/OFF and to control handMore Product Information