Solaris 4000 R Large Refrigerated Incubating Benchtop Orbital Shakers
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SK4002 SK4002CPKG SK4002MPKG
444-1439EA 8568 EUR
444-1439 171-1520 BARNSK4002MPKG
Solaris 4000 R Large Refrigerated Incubating Benchtop Orbital Shakers
Schudapparaten Schudincubatoren
Solaris incubated refrigerated orbital shakers feature Peltier chillers, units have a compact footprint, are designed to fit on the benchtop, user programmability for easy setup and reproducibility plus connectivity for easy monitoring.
  • Exceptional reliability with heavy-duty drive mechanism
  • Ability to create, download, and share up to 99 programs across multiple units
  • USB and ethernet connections for monitoring and data output
  • Refrigerant-free, environmentally friendly Peltier cooling, which uses 58% less energy compared to older models with traditional
More Product Information