EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride)

Leverancier: G Bioscienes
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: EMCH

BC15
BC15EA 771 EUR
BC15
EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride)
EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride)

N-(ε-Maleimidocaproic acid) hydrazide Heterobifunctional Cross Linker.

Formule: C₁₀H₁₅N₃O₃·ClH
MW: 261.705 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD07363434
CAS nummer: 175290-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03785808e11a7f5bb3d47e048cb7a7a0f70d9c6b&vid=03785808e11a7f5bb3d47e048cb7a7a0f70d9c6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Spacer Arm (Å) 11.8
Reactive Toward Sulfhydryl + Carbohydrate
Membrane Permeable Not tested
Water Soluble No
Cleavable/ Reversible No

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR