Guanidiniumchloride

Leverancier: G Bioscienes
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

BC88 BC85 RC-057
BC88EA 463 EUR
BC88 BC85 RC-057
Guanidiniumchloride
Guanidiniumchloride

A selection of strong chemicals to aid in the denaturation of proteins. Denaturants, or chaotropes, disrupt water interactions resulting in the solubilization of hydrophobic proteins and peptides. The chaotropes also act on all proteins to unfold them and alter their three-dimensional structure.


Available as a powder or 8M solution

Formule: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Smeltpunt: 185
Dichtheid: 1.354 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013026
CAS nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR