Racks for cryogenic vials, TruCool®
Leverancier: BIOCISION
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BCS-222
BCISBCS-222EA 350 EUR
BCISBCS-222
Racks for cryogenic vials, TruCool®
Rekken Rekken voor cryovials
These polypropylene cryogenic racks feature a locking mechanism that allow one handed opening for self-standing cryogenic vials.
  • Accommodates both round bottom and self-standing vial formats
  • Racks have A to J row and 1 to 5 column indexing for easy organization
  • Stackable when not in use
Multicolour pack contains one each of blue, green, orange, purple and pink racks.