Antibody diluent clear
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BD09-125 BD09-500 BD09-999 S/BD09-20
BD09-125EA 57.8 EUR
BD09-125 BD09-500 BD09-999 IMMLS/BD09-20
Antibody diluent clear
Reagentia voor elektroforese Western Blotting Reagents
Universal IHC Blocking/Diluent is specially formulated to stabilise antibodies and eliminate backgrounds encountered in immunohistochemistry staining procedures. The diluent contains complex proteins and non-protein mixtures which eliminate histostaining background by blocking Fc receptors and prevents non-specific protein-protein interactions.
  • High-quality diluent solution for IHC apllications
Universal IHC Blocking/Diluent can also be used as blocking applied before primary antibodies. If primary antibodies are diluted in this diluent, a blocking step before primary antibodies can generally be ommited. The blocking/diluent solution is supplied in ready-to-use format. It should not be diluted to be effective. The diluent contains no azide, therefore, it can be used to dilute and stabilise HRP-conjugates as well as any other solution that need and stabilising carrier protein, such as post-antibody blocking and poly-AP conjugates.
Certificaten: Meets CE/IVD standards
Bestelinformatie: Colour version also available (see IMMLS/UD09-20 and similar products)
Leveringsinformatie: Shipped with blue ice. Store the products between 2 to 8°C.
Opgelet: Irritating to eyes, respiratory systems and skin
Order Now
Specificaties
Environmentally Preferable